Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Caffeoyl O-methyltyramine | 189307-47-9 | 25 MG
SDP

Catalog No. 5000421680
Encompass_Preferred

N-Caffeoyl O-methyltyramine is an alkaloid isolated from Cuscuta reflexa that demonstrates strong inhibitory activity against α-glucosidase, with an IC50 of 103.58 μM. This product is intended for research use only.

  • Isolated from Cuscuta reflexa
  • Exhibits strong inhibitory activity against α-glucosidase
  • Molecular formula: C18H19NO4
  • Molecular weight: 313.35
  • Purity: 98.52%
  • Appearance: Solid, white to off-white

Catalog No. 50-004-21680 Supplier Medchemexpress LLC Supplier No. 25MGHYN7203
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Caffeoyl O-methyltyramine is an alkaloid isolated from Cuscuta reflexa that demonstrates strong inhibitory activity against α-glucosidase, with an IC50 of 103.58 μM. This product is intended for research use only.

  • Isolated from Cuscuta reflexa
  • Exhibits strong inhibitory activity against α-glucosidase
  • Molecular formula: C18H19NO4
  • Molecular weight: 313.35
  • Purity: 98.52%
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.