Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Caffeoyl O-methyltyramine | 189307-47-9 | 5 MG
SDP

Catalog No. 5000418978
Encompass_Preferred

N-Caffeoyl O-methyltyramine is an alkaloid isolated from *Cuscuta reflexa* known for its strong inhibitory activity against α-glucosidase. This compound, with a molecular weight of 313.35 and a molecular formula of C18H19NO4, is typically supplied as a solid.

  • Inhibits α-glucosidase with an IC50 of 103.58 μM
  • Molecular weight: 313.35
  • Molecular formula: C18H19NO4
  • Appearance: Solid, white to off-white
  • Classified as an alkaloid, phenol, and polyphenol
  • Derived from plants, specifically *Fissistigma oldhamii* (Hemsl.) Merr.
  • Targets glycosidase
  • Pathway: Metabolic enzyme/protease

Catalog No. 50-004-18978 Supplier Medchemexpress LLC Supplier No. HYN72035MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Caffeoyl O-methyltyramine is an alkaloid isolated from *Cuscuta reflexa* known for its strong inhibitory activity against α-glucosidase. This compound, with a molecular weight of 313.35 and a molecular formula of C18H19NO4, is typically supplied as a solid.

  • Inhibits α-glucosidase with an IC50 of 103.58 μM
  • Molecular weight: 313.35
  • Molecular formula: C18H19NO4
  • Appearance: Solid, white to off-white
  • Classified as an alkaloid, phenol, and polyphenol
  • Derived from plants, specifically *Fissistigma oldhamii* (Hemsl.) Merr.
  • Targets glycosidase
  • Pathway: Metabolic enzyme/protease

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.