Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Caffeoyl O-methyltyramine | 189307-47-9 | 50 MG
SDP

Catalog No. 5000417269
Encompass_Preferred

N-Caffeoyl O-methyltyramine is a class of alkaloid isolated from Cuscuta reflexa with strong inhibitory activity against α-glucosidase. It is presented as a white to off-white solid with a purity of 98.52%.

  • Strong inhibitory activity against α-glucosidase (IC50 of 103.58 μM)
  • Isolated from *Cuscuta reflexa*
  • White to off-white solid appearance
  • Purity of 98.52%
  • Soluble in DMSO (250 mg/mL)

Catalog No. 50-004-17269 Supplier Medchemexpress LLC Supplier No. HYN720350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Caffeoyl O-methyltyramine is a class of alkaloid isolated from Cuscuta reflexa with strong inhibitory activity against α-glucosidase. It is presented as a white to off-white solid with a purity of 98.52%.

  • Strong inhibitory activity against α-glucosidase (IC50 of 103.58 μM)
  • Isolated from *Cuscuta reflexa*
  • White to off-white solid appearance
  • Purity of 98.52%
  • Soluble in DMSO (250 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.