Learn More
Selleck Chemical LLC N-Caffeoyl O-methyltyramine
SDP
Supplier:  Selleck Chemical LLC E04775MG
Encompass_Preferred
N-Caffeoyl O-methyltyramine is a class of alkaloid isolated from Cuscuta reflexa with strong inhibitory activity against -glucosidase with IC50 of 103 58 M
Catalog No. 50-313-7077