Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Caffeoylputrescine,(E)- | 29554-26-5 | 5 MG
SDP

Catalog No. 5000418905
Encompass_Preferred

N-Caffeoylputrescine,(E)- is a caffeic acid amide found in tobacco plants (Nicotiana tabacum L.). It is intended for research use only, with a purity of 99.27%. This product has a molecular weight of 250.29 and the chemical formula C13H18N2O3. It appears as a white to yellow solid and is initially sourced from various plants, including Nicotiana tabacum L.

  • Purity of 99.27%
  • Molecular weight of 250.29
  • Chemical formula C13H18N2O3
  • Appears as a white to yellow solid
  • Initially sourced from plants including Nicotiana tabacum L.
  • Recommended storage at 4°C, protected from light

Catalog No. 50-004-18905 Supplier Medchemexpress LLC Supplier No. HYN60855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Caffeoylputrescine,(E)- is a caffeic acid amide found in tobacco plants (Nicotiana tabacum L.). It is intended for research use only, with a purity of 99.27%. This product has a molecular weight of 250.29 and the chemical formula C13H18N2O3. It appears as a white to yellow solid and is initially sourced from various plants, including Nicotiana tabacum L.

  • Purity of 99.27%
  • Molecular weight of 250.29
  • Chemical formula C13H18N2O3
  • Appears as a white to yellow solid
  • Initially sourced from plants including Nicotiana tabacum L.
  • Recommended storage at 4°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.