Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Capric Acid, MP Biomedicals

Catalog No. ICN209804 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209804 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209804 Supplier MP Biomedicals, Inc Supplier No. 0520980450
Add to Cart
Add to Cart

Chemical Identifiers

CAS 334-48-5
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
InChI Key GHVNFZFCNZKVNT-UHFFFAOYSA-N
Synonym capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
PubChem CID 2969
ChEBI CHEBI:30813
IUPAC Name decanoic acid
SMILES CCCCCCCCCC(=O)O

Specifications

Quantity 100 g
Formula Weight 172.27
Chemical Name or Material n-Capric Acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.