Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Carbamimidoyl-N-(methyl-d3)glycine-2,2-d2 hydrate | 99.9% | 154.18 g/mol | C4H6D5N3O3 | 10 MG
SDP

Catalog No. 5000300495
Encompass_Preferred

Creatine-d5 monohydrate is a deuterium-labeled isotopic standard of creatine monohydrate designed for research and analytical applications. It is used as an internal standard in mass spectrometry and other bioanalytical assays to improve quantitation accuracy and traceability in metabolic and pharmacokinetic studies.

  • High isotopic enrichment for reliable mass-differentiation.
  • High chemical purity (99.93%).
  • Molecular formula: C4H6D5N3O3.
  • Molecular weight: 154.18 g/mol.
  • Provided with certificate of analysis and datasheet.
  • Available in small pack sizes suitable for analytical use.
  • Suitable as an internal standard for LC-MS applications.

Catalog No. 50-003-00495 Supplier Medchemexpress LLC Supplier No. HYW017462S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Creatine-d5 monohydrate is a deuterium-labeled isotopic standard of creatine monohydrate designed for research and analytical applications. It is used as an internal standard in mass spectrometry and other bioanalytical assays to improve quantitation accuracy and traceability in metabolic and pharmacokinetic studies.

  • High isotopic enrichment for reliable mass-differentiation.
  • High chemical purity (99.93%).
  • Molecular formula: C4H6D5N3O3.
  • Molecular weight: 154.18 g/mol.
  • Provided with certificate of analysis and datasheet.
  • Available in small pack sizes suitable for analytical use.
  • Suitable as an internal standard for LC-MS applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.