Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Carbamoyl-DL-aspartic acid | 923-37-5 | 1 G
SDP

Catalog No. 5000339255
Encompass_Preferred

N-Carbamoyl-DL-aspartic acid, also known as Ureidosuccinic acid, is a precursor of nucleic acid pyrimidines. It has demonstrated antitumor activities and is considered an endogenous metabolite, playing a role in metabolic enzyme/protease pathways.

  • Precursor of nucleic acid pyrimidines
  • Exhibits antitumor activities
  • Effective in murine Lewis lung carcinoma models
  • Reverses toxicity and antitumor activity of N-(phosphonacetyl)-L-aspartate
  • Functions as a precursor of nucleic acid pyrimidines in normal and tumor-bearing mice
  • Soluble in DMSO for in vitro studies
  • Can be formulated for in vivo studies using various solvent systems
  • Recommended storage for powder is -20°C for 3 years or 4°C for 2 years
  • Recommended storage in solvent is -80°C for 2 years or -20°C for 1 year
  • For research use only

Catalog No. 50-003-39255 Supplier Medchemexpress LLC Supplier No. HY1284251G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Carbamoyl-DL-aspartic acid, also known as Ureidosuccinic acid, is a precursor of nucleic acid pyrimidines. It has demonstrated antitumor activities and is considered an endogenous metabolite, playing a role in metabolic enzyme/protease pathways.

  • Precursor of nucleic acid pyrimidines
  • Exhibits antitumor activities
  • Effective in murine Lewis lung carcinoma models
  • Reverses toxicity and antitumor activity of N-(phosphonacetyl)-L-aspartate
  • Functions as a precursor of nucleic acid pyrimidines in normal and tumor-bearing mice
  • Soluble in DMSO for in vitro studies
  • Can be formulated for in vivo studies using various solvent systems
  • Recommended storage for powder is -20°C for 3 years or 4°C for 2 years
  • Recommended storage in solvent is -80°C for 2 years or -20°C for 1 year
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.