Learn More
Medchemexpress LLC N-Carbamoyl-DL-aspartic acid | 923-37-5 | 99.9% | 1 ML
SDP
Catalog No. 5000341201
Encompass_Preferred
N-Carbamoyl-DL-aspartic acid, also known as Ureidosuccinic acid, is a chemical compound that serves as a precursor for nucleic acid pyrimidines. It has demonstrated antitumor activities.
  • Functions as a precursor of nucleic acid pyrimidines.
  • Exhibits antitumor properties.
  • Shown to inhibit antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA) in in vivo studies.
Catalog No. 50-003-41201 Supplier Medchemexpress LLC Supplier No. HY12842510MM/1ML
N-Carbamoyl-DL-aspartic acid, also known as Ureidosuccinic acid, is a chemical compound that serves as a precursor for nucleic acid pyrimidines. It has demonstrated antitumor activities.
  • Functions as a precursor of nucleic acid pyrimidines.
  • Exhibits antitumor properties.
  • Shown to inhibit antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA) in in vivo studies.