Learn More
Medchemexpress LLC N-Carbamoyl-DL-aspartic acid | 923-37-5 | 99.9% | 10 MG
SDP
Catalog No. 5000339108
Encompass_Preferred
N-Carbamoyl-DL-aspartic acid, also known as Ureidosuccinic acid, is a precursor of nucleic acid pyrimidines with reported antitumor activities. It has been shown to inhibit antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA) in a murine Lewis lung carcinoma model.
  • Precursor of nucleic acid pyrimidines
  • Exhibits antitumor activities
  • Identified for use as laboratory chemical
  • Stable under recommended storage conditions
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-003-39108 Supplier Medchemexpress LLC Supplier No. HY128425R10MG
N-Carbamoyl-DL-aspartic acid, also known as Ureidosuccinic acid, is a precursor of nucleic acid pyrimidines with reported antitumor activities. It has been shown to inhibit antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA) in a murine Lewis lung carcinoma model.
  • Precursor of nucleic acid pyrimidines
  • Exhibits antitumor activities
  • Identified for use as laboratory chemical
  • Stable under recommended storage conditions
  • White to off-white solid appearance
  • For research use only