Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Carbamoyl-DL-aspartic acid | 923-37-5 | 99.9% | 100 MG
SDP

Catalog No. 5000339778
Encompass_Preferred

N-Carbamoyl-DL-aspartic acid (Ureidosuccinic acid) is a precursor of nucleic acid pyrimidines and possesses antitumor activities. It has been shown to inhibit antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA) in a murine Lewis lung carcinoma model.

  • Precursor of nucleic acid pyrimidines
  • Exhibits antitumor activities
  • Inhibits antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA)
  • Molecular weight: 176.13
  • Formula: C5H8N2O5
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-003-39778 Supplier Medchemexpress LLC Supplier No. HY128425R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

N-Carbamoyl-DL-aspartic acid (Ureidosuccinic acid) is a precursor of nucleic acid pyrimidines and possesses antitumor activities. It has been shown to inhibit antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA) in a murine Lewis lung carcinoma model.

  • Precursor of nucleic acid pyrimidines
  • Exhibits antitumor activities
  • Inhibits antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA)
  • Molecular weight: 176.13
  • Formula: C5H8N2O5
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.