Learn More
Medchemexpress LLC N-Carbamoyl-DL-aspartic acid | 923-37-5 | 99.9% | 5 G
SDP
Catalog No. 5000339251
Encompass_Preferred
N-Carbamoyl-DL-aspartic acid, also known as Ureidosuccinic acid, is a precursor of nucleic acid pyrimidines and exhibits antitumor activities. It is intended for research use only and not for patient administration.
  • Precursor of nucleic acid pyrimidines
  • Exhibits antitumor activities
  • Inhibits antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA)
  • Endogenous metabolite
  • Inhibitor
Catalog No. 50-003-39251 Supplier Medchemexpress LLC Supplier No. HY1284255G
N-Carbamoyl-DL-aspartic acid, also known as Ureidosuccinic acid, is a precursor of nucleic acid pyrimidines and exhibits antitumor activities. It is intended for research use only and not for patient administration.
  • Precursor of nucleic acid pyrimidines
  • Exhibits antitumor activities
  • Inhibits antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA)
  • Endogenous metabolite
  • Inhibitor