Learn More
Medchemexpress LLC N-Carbamoyl-DL-aspartic acid | 923-37-5 | 99.9% | 500 MG
SDP
Catalog No. 5000339265
Encompass_Preferred
N-Carbamoyl-DL-aspartic acid (Ureidosuccinic acid) is an endogenous metabolite and a precursor of nucleic acid pyrimidines. This compound is known for its antitumor activities.
  • Precursor of nucleic acid pyrimidines.
  • Exhibits antitumor activities.
  • Demonstrates antitumor activity in a murine Lewis lung carcinoma model.
  • Inhibits antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA) in animal models.
  • Intended for research use only.
Catalog No. 50-003-39265 Supplier Medchemexpress LLC Supplier No. HY128425500MG
Add to Cart
Add to Cart
N-Carbamoyl-DL-aspartic acid (Ureidosuccinic acid) is an endogenous metabolite and a precursor of nucleic acid pyrimidines. This compound is known for its antitumor activities.
  • Precursor of nucleic acid pyrimidines.
  • Exhibits antitumor activities.
  • Demonstrates antitumor activity in a murine Lewis lung carcinoma model.
  • Inhibits antitumor activity and toxicity induced by N-phosphonacetyl-L-aspartate (PALA) in animal models.
  • Intended for research use only.