Learn More
Medchemexpress LLC N-carbamoyl-DL-aspartic acid | 923-37-5 | 99.92% | 50 MG
SDP
Catalog No. 5000341204
Encompass_Preferred
N-Carbamoyl-DL-aspartic acid (Ureidosuccinic acid) is the analytical standard of N-Carbamoyl-DL-aspartic acid. This product is intended for research and analytical applications. It is a precursor of nucleic acid pyrimidines and has antitumor activities. The compound is a reference standard commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Used as a reference standard assay
  • Intended for research and analytical applications
  • Involved in fundamental biological processes as a precursor of nucleic acid pyrimidines
  • Possesses potential therapeutic interest due to antitumor activities
  • Commonly used in HPLC, GC, and MS research experiments
Catalog No. 50-003-41204 Supplier Medchemexpress LLC Supplier No. HY128425R50MG
N-Carbamoyl-DL-aspartic acid (Ureidosuccinic acid) is the analytical standard of N-Carbamoyl-DL-aspartic acid. This product is intended for research and analytical applications. It is a precursor of nucleic acid pyrimidines and has antitumor activities. The compound is a reference standard commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Used as a reference standard assay
  • Intended for research and analytical applications
  • Involved in fundamental biological processes as a precursor of nucleic acid pyrimidines
  • Possesses potential therapeutic interest due to antitumor activities
  • Commonly used in HPLC, GC, and MS research experiments