Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbamoyl-DL-aspartic Acid 98.0+%, TCI America™
SDP

Catalog No. C00291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C00291G Supplier TCI America Supplier No. C00291G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 923-37-5
Molecular Formula C5H8N2O5
Molecular Weight (g/mol) 176.128
MDL Number MFCD00042822
InChI Key HLKXYZVTANABHZ-UHFFFAOYSA-N
Synonym ureidosuccinic acid, 2-ureidosuccinic acid, n-carbamoyl-dl-aspartic acid, n-aminocarbonyl-dl-aspartic acid, dl-aspartic acid, n-aminocarbonyl, 2-carbamoylamino butanedioic acid, n-carbamoylaspartic acid, carbamoyl-dl-aspartic acid, carbamylaspartic acid, 2-ureidobutanedioic acid
PubChem CID 279
ChEBI CHEBI:64850
IUPAC Name 2-(carbamoylamino)butanedioic acid
SMILES C(C(C(=O)O)NC(=O)N)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 176.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbamoyl-DL-aspartic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.