Learn More
Medchemexpress LLC N-Carbamoyl-L-aspartic acid | 13184-27-5 | >99.8% | 10 MG
SDP
Catalog No. 5000339252
Encompass_Preferred
N-Carbamoyl-L-aspartic acid, also known as L-Ureidosuccinic acid, is an essential intermediate in the pyrimidine metabolism pathway. This compound plays a crucial role as a precursor for nucleic acid pyrimidines and is frequently utilized in biochemical research to assess the enzymatic activity of dihydroorotase (DHOase). It is available as a white to off-white solid.
  • Intermediate of pyrimidine metabolism
  • Used to measure dihydroorotase (DHOase) enzyme activity
  • White to off-white solid
  • High purity confirmed by LCMS
Catalog No. 50-003-39252 Supplier Medchemexpress LLC Supplier No. HY128425A10MG
N-Carbamoyl-L-aspartic acid, also known as L-Ureidosuccinic acid, is an essential intermediate in the pyrimidine metabolism pathway. This compound plays a crucial role as a precursor for nucleic acid pyrimidines and is frequently utilized in biochemical research to assess the enzymatic activity of dihydroorotase (DHOase). It is available as a white to off-white solid.
  • Intermediate of pyrimidine metabolism
  • Used to measure dihydroorotase (DHOase) enzyme activity
  • White to off-white solid
  • High purity confirmed by LCMS