Learn More
Medchemexpress LLC N-carbamoyl-L-aspartic acid | 13184-27-5 | MFCD01664827 | 98.0% | 176.13 g/mol | C5H8N2O5 | 5 MG
SDP
Catalog No. 5000264350
Encompass_Preferred
N-Carbamoyl-L-aspartic acid is the L-enantiomer of ureidosuccinic acid and an intermediate in pyrimidine biosynthesis. It is supplied as a research-grade reagent for biochemical and enzymatic studies, including measurement of dihydroorotase activity. Analytical documentation and spectral data are provided to support experimental use.
  • Intermediate in pyrimidine metabolism
  • Suitable for enzyme activity assays such as dihydroorotase
  • High reported purity (98.0%)
  • Available in small research pack sizes
  • Analytical documents available: data sheet, COA, SDS, NMR, MS
Catalog No. 50-002-64350 Supplier Medchemexpress LLC Supplier No. HY128425A5MG
N-Carbamoyl-L-aspartic acid is the L-enantiomer of ureidosuccinic acid and an intermediate in pyrimidine biosynthesis. It is supplied as a research-grade reagent for biochemical and enzymatic studies, including measurement of dihydroorotase activity. Analytical documentation and spectral data are provided to support experimental use.
  • Intermediate in pyrimidine metabolism
  • Suitable for enzyme activity assays such as dihydroorotase
  • High reported purity (98.0%)
  • Available in small research pack sizes
  • Analytical documents available: data sheet, COA, SDS, NMR, MS