Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Carbamoyl-L-cysteine

Catalog No. CDS00644 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS00644 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS00644 Supplier Sigma Aldrich Supplier No. CDS00644250MG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,5-bis(trifluoromethyl)aniline was used in the synthesis of:
  • N-[2-hydroxy-1-naphthylidene]-3, 5-bis(trifluoromethyl)aniline, schiffαs base
  • 5,7-bis(trifluoromethyl)aniline
  • N-1-phenylethyl-3,5-bis(trifluoromethyl)aniline via titanium-catalyzed hydroamination reaction
  • N,nα-bis[3,5-bis(tri-fluoromethyl)phenyl]thiourea, organocatalyst

Specifications

CAS 24583-23-1
Molecular Formula C4H8N2O3S
Linear Formula C4H8N2O3S
MDL Number MFCD00152026
Quantity 10 g
Molecular Weight (g/mol) 164.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.