Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Carbobenzoxy)aminoacetonitrile 95.0+%, TCI America™
SDP

Catalog No. C14395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14395G 5 g
C143925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14395G Supplier TCI America Supplier No. C14395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3589-41-1
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD00033175
InChI Key DVUXKEFDAGQPQU-UHFFFAOYSA-N
Synonym Benzyl Cyanomethylcarbamate, N-Cbz-aminoacetonitrile, Cyanomethylcarbamic Acid Benzyl Ester
PubChem CID 244143
IUPAC Name benzyl N-(cyanomethyl)carbamate
SMILES O=C(NCC#N)OCC1=CC=CC=C1

Specifications

Melting Point 59°C
Quantity 5 g
Formula Weight 190.20
Percent Purity ≥95.0% (N)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material N-(Carbobenzoxy)aminoacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.