Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-D-aspartic Acid 98.0+%, TCI America™
SDP

Catalog No. C06895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06895G 5 g
C06891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C06895G Supplier TCI America Supplier No. C06895G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78663-07-7
Molecular Formula C12H13NO6
Molecular Weight (g/mol) 267.237
MDL Number MFCD00063182
InChI Key XYXYXSKSTZAEJW-SECBINFHSA-N
Synonym z-d-asp-oh, n-benzyloxycarbonyl-d-aspartic acid, z-d-aspartic acid, cbz-d-aspartic acid, n-cbz-d-aspartic acid, r-2-benzyloxy carbonyl amino succinic acid, cbz-d-asp-oh, n-carbobenzoxy-d-aspartic acid, r-2-benzyloxycarbonylamino succinic acid, 2r-2-benzyloxy carbonyl amino butanedioic acid
PubChem CID 5289152
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O

Specifications

Melting Point 117°C
Color White
Quantity 5 g
Formula Weight 267.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-aspartic Acid
EINECSNumber (9)-1549
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.