Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-carbobenzoxy-D-homoserine | 41088-85-1 | MFCD12545887 | 253.25 g/mol | C12H15NO5 | 5 G
SDP

Catalog No. 5000299689
Encompass_Preferred

N-Carbobenzoxy-D-homoserine (N-(Benzyloxycarbonyl)-D-homoserine) is the D-enantiomer of homoserine bearing an N-carbobenzoxy (Cbz) protecting group. Supplied as a powder, it is used as a synthetic intermediate in organic and peptide synthesis, including preparation of antibody-drug conjugates and other constructs that require temporary N-protection.

  • Powdered synthetic intermediate for organic synthesis.
  • Contains N-carbobenzoxy (Cbz) protecting group for the amino function.
  • Suitable for peptide synthesis and antibody-drug conjugate payload or linker preparation.
  • Molecular formula C12H15NO5; molecular weight 253.25 g/mol.
  • Recommended storage: powder -20°C for long term, 4°C for short term.

Catalog No. 50-002-99689 Supplier Medchemexpress LLC Supplier No. HYW025723A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Carbobenzoxy-D-homoserine (N-(Benzyloxycarbonyl)-D-homoserine) is the D-enantiomer of homoserine bearing an N-carbobenzoxy (Cbz) protecting group. Supplied as a powder, it is used as a synthetic intermediate in organic and peptide synthesis, including preparation of antibody-drug conjugates and other constructs that require temporary N-protection.

  • Powdered synthetic intermediate for organic synthesis.
  • Contains N-carbobenzoxy (Cbz) protecting group for the amino function.
  • Suitable for peptide synthesis and antibody-drug conjugate payload or linker preparation.
  • Molecular formula C12H15NO5; molecular weight 253.25 g/mol.
  • Recommended storage: powder -20°C for long term, 4°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.