Learn More
Medchemexpress LLC N-carbobenzoxy-D-homoserine (N-(benzyloxycarbonyl)-D-homoserine) | 41088-85-1 | MFCD12545887 | >98.0% | 253.25 g·mol⁻¹ | C12H15NO5 | 10 G
SDP
Catalog No. 5000301246
Encompass_Preferred
N-Carbobenzoxy-D-homoserine (N-(Benzyloxycarbonyl)-D-homoserine) is a Cbz-protected D-homoserine used as a synthetic intermediate and building block in medicinal chemistry. It is employed in the preparation of antibody-drug conjugates and peptides, providing a protected amino acid handle for selective coupling and downstream functionalization.
  • Cbz-protected amino acid for selective N-protection.
  • Suitable intermediate for antibody-drug conjugate synthesis.
  • Compatible with peptide and amide bond formation chemistries.
  • Stable as a powder with defined low-temperature storage recommendations.
  • Characterized by a known molecular formula and molecular weight for analytical use.
Catalog No. 50-003-01246 Supplier Medchemexpress LLC Supplier No. HYW025723A10G
N-Carbobenzoxy-D-homoserine (N-(Benzyloxycarbonyl)-D-homoserine) is a Cbz-protected D-homoserine used as a synthetic intermediate and building block in medicinal chemistry. It is employed in the preparation of antibody-drug conjugates and peptides, providing a protected amino acid handle for selective coupling and downstream functionalization.
  • Cbz-protected amino acid for selective N-protection.
  • Suitable intermediate for antibody-drug conjugate synthesis.
  • Compatible with peptide and amide bond formation chemistries.
  • Stable as a powder with defined low-temperature storage recommendations.
  • Characterized by a known molecular formula and molecular weight for analytical use.