Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-D-leucine 97.0+%, TCI America™
SDP

Catalog No. C28395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28395G 5 g
C283925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28395G Supplier TCI America Supplier No. C28395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28862-79-5
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00066068
InChI Key USPFMEKVPDBMCG-GFCCVEGCSA-N
Synonym z-d-leu-oh, n-cbz-d-leucine, cbz-d-leu-oh, z-d-leucine, n-benzyloxycarbonyl-d-leucine, r-2-benzyloxy carbonyl amino-4-methylpentanoic acid, cbz-d-leucine, n-alpha-benzyloxycarbonyl-d-leucine, 2r-4-methyl-2-phenylmethoxycarbonylamino pentanoic acid, 2r-2-benzyloxy carbonyl amino-4-methylpentanoic acid
PubChem CID 7000099
IUPAC Name (2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid
SMILES CC(C)C[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Color Colorless
Quantity 5 g
Formula Weight 265.31
Percent Purity ≥97.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material N-Carbobenzoxy-D-leucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.