Learn More
Medchemexpress LLC N-carbobenzoxy-D-serine methyl ester | 93204-36-5 | MFCD00799501 | 253.25 g·mol⁻1 | C12H15NO5 | 10 G
SDP
Catalog No. 5000210015
Encompass_Preferred
Methyl ((benzyloxy)carbonyl)-D-serinate (N-carbobenzoxy-D-serine methyl ester) is a protected D-serine derivative used as an amino acid building block and intermediate in organic and peptide synthesis. It is supplied as a powder for research applications.
  • Protected D-serine methyl ester for use in synthetic chemistry
  • Useful as a building block in peptide synthesis and derivatization
  • Molecular formula C12H15NO5; molecular weight 253.25 g·mol⁻1
  • Identified by CAS number 93204-36-5 for unambiguous substance reference
  • Store powder at -20°C for long-term stability, or 4°C for shorter-term storage
Catalog No. 50-002-10015 Supplier Medchemexpress LLC Supplier No. HYW05231010G
Methyl ((benzyloxy)carbonyl)-D-serinate (N-carbobenzoxy-D-serine methyl ester) is a protected D-serine derivative used as an amino acid building block and intermediate in organic and peptide synthesis. It is supplied as a powder for research applications.
  • Protected D-serine methyl ester for use in synthetic chemistry
  • Useful as a building block in peptide synthesis and derivatization
  • Molecular formula C12H15NO5; molecular weight 253.25 g·mol⁻1
  • Identified by CAS number 93204-36-5 for unambiguous substance reference
  • Store powder at -20°C for long-term stability, or 4°C for shorter-term storage