Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-D-serine 98.0+%, TCI America™
SDP

Catalog No. C21375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21375G 5 g
C21371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21375G Supplier TCI America Supplier No. C21375G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6081-61-4
Molecular Formula C11H13NO5
Molecular Weight (g/mol) 239.227
MDL Number MFCD00063144
InChI Key GNIDSOFZAKMQAO-SECBINFHSA-N
Synonym z-d-ser-oh, n-cbz-d-serine, cbz-d-ser-oh, z-d-serine, n-carbobenzoxy-d-serine, n-carbobenzyloxy-d-serine, r-2-benzyloxy carbonyl amino-3-hydroxypropanoic acid, cbz-d-serine, n-benzyloxycarbonyl-d-serine, benzyloxycarbonyl-d-serine
PubChem CID 489184
IUPAC Name (2R)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CO)C(=O)O

Specifications

Melting Point 118°C
Color White
Quantity 5 g
Formula Weight 239.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.