Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-D-tyrosine 98.0+%, TCI America™
SDP

Catalog No. C21241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21241G 1 g
C21245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21241G Supplier TCI America Supplier No. C21241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64205-12-5
Molecular Formula C17H17NO5
Molecular Weight (g/mol) 315.325
MDL Number MFCD00063031
InChI Key MCRMUCXATQAAMN-OAHLLOKOSA-N
Synonym N-Cbz-D-tyrosine, Z-D-Tyr-OH
PubChem CID 736160
IUPAC Name (2R)-3-(4-hydroxyphenyl)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC2=CC=C(C=C2)O)C(=O)O

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 1 g
Formula Weight 315.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.