Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-DL-alanine 99.0+%, TCI America™
SDP

Catalog No. C063210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C063210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C063210G Supplier TCI America Supplier No. C063210G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4132-86-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00063125
InChI Key TYRGLVWXHJRKMT-UHFFFAOYNA-N
Synonym z-dl-ala-oh, dl-cbz-alanine, 2-benzyloxy carbonyl amino propanoic acid, n-cbz-dl-alanine, n-carbobenzoxy-dl-alanine, n-benzyloxy carbonyl alanine, carbobenzyloxy-dl-alanine, n-benzyloxycarbonyl-dl-alanine, n-carbobenzoxyalanine, carbobenzoxy-dl-alanine
PubChem CID 95657
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}propanoic acid
SMILES CC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 114°C
Color White
Quantity 10 g
Formula Weight 223.23
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.