Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-DL-serine 98.0+%, TCI America™
SDP

Catalog No. C063710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C063710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C063710G Supplier TCI America Supplier No. C063710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2768-56-1
Molecular Formula C11H13NO5
Molecular Weight (g/mol) 239.227
MDL Number MFCD00063143
InChI Key GNIDSOFZAKMQAO-UHFFFAOYSA-N
Synonym n-cbz-dl-serine, z-dl-ser-oh, n-carbobenzoxy-dl-serine, n-carbobenzyloxy-dl-serine, n-benzyloxycarbonyl-dl-serine, 2-benzyloxy carbonyl amino-3-hydroxypropanoic acid, carbobenzyloxy-d,l-serine, dl-serine, n-phenylmethoxy carbonyl, serine, n-benzyl ester, dl
PubChem CID 76007
IUPAC Name 3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CO)C(=O)O

Specifications

Melting Point 125°C
Color White
Quantity 10 g
Formula Weight 239.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.