Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-DL-tryptophan 97.0+%, TCI America™
SDP

Catalog No. C06411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06411G 1 g
C06415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C06411G Supplier TCI America Supplier No. C06411G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13058-16-7
Molecular Formula C19H18N2O4
Molecular Weight (g/mol) 338.36
MDL Number MFCD00069707
InChI Key AHYFYYVVAXRMKB-UHFFFAOYNA-N
Synonym N-Cbz-DL-tryptophan, Z-DL-Trp-OH
PubChem CID 98198
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)C(CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 170°C
Quantity 1 g
Formula Weight 338.36
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.