Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-DL-valine 99.0+%, TCI America™
SDP

Catalog No. C06341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06341G 1 g
C063410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C06341G Supplier TCI America Supplier No. C06341G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3588-63-4
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00065127
InChI Key CANZBRDGRHNSGZ-UHFFFAOYSA-N
Synonym n-carbobenzoxy-dl-valine, z-dl-val-oh, n-cbz-dl-valine, 2-benzyloxy carbonyl amino-3-methylbutanoic acid, n-carbobenzoyl-dl-valine, n-benzyloxy carbonyl valine, n-benzyloxycarbonyl-dl-valine, n-carbobenzyl-oxy-dl-valine, dl-valine, n-phenylmethoxy carbonyl
PubChem CID 97743
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
SMILES CC(C)C(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 78°C
Color White
Quantity 1 g
Formula Weight 251.28
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-valine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.