Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-glutamic Acid 98.0+%, TCI America™
SDP

Catalog No. C073425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C073425G 25 g
C0734100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C073425G Supplier TCI America Supplier No. C073425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1155-62-0
Molecular Formula C13H15NO6
Molecular Weight (g/mol) 281.264
MDL Number MFCD00002801
InChI Key PVFCXMDXBIEMQG-JTQLQIEISA-N
Synonym n-cbz-l-glutamic acid, z-glu-oh, n-benzyloxycarbonyl-l-glutamic acid, carbobenzoxy-l-glutamic acid, n-carbobenzoxy-l-glutamic acid, s-2-benzyloxy carbonyl amino pentanedioic acid, n-benzyloxycarbonylglutamic acid, n-carbobenzyloxy-l-glutamic acid, z-l-glu-oh
PubChem CID 70866
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 120°C
Color White
Quantity 25 g
Formula Weight 281.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-glutamic Acid
EINECSNumber (9)-2100
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.