Learn More
Medchemexpress LLC N-carbobenzoxy-L-homoserine | 35677-88-4 | MFCD00056718 | >98.0% | 253.25 g·mol⁻¹ | C12H15NO5 | 1 G
SDP
Catalog No. 5000301641
Encompass_Preferred
N-Carbobenzoxy-L-homoserine is the Cbz (benzyloxycarbonyl)-protected derivative of L-homoserine used as a synthetic building block in peptide and small-molecule synthesis. It is supplied as a white to off-white solid (CAS 35677-88-4) and provides temporary protection of the amino group during multi-step syntheses.
  • Protects amino group in peptide and small-molecule synthesis.
  • Suitable as a Cbz-protected L-homoserine building block.
  • Supplied as a white to off-white solid for easy handling.
  • Melting point around 100-101 °C for characterization.
  • Available in small laboratory packaging sizes for research use.
Catalog No. 50-003-01641 Supplier Medchemexpress LLC Supplier No. HYW0257231G
N-Carbobenzoxy-L-homoserine is the Cbz (benzyloxycarbonyl)-protected derivative of L-homoserine used as a synthetic building block in peptide and small-molecule synthesis. It is supplied as a white to off-white solid (CAS 35677-88-4) and provides temporary protection of the amino group during multi-step syntheses.
  • Protects amino group in peptide and small-molecule synthesis.
  • Suitable as a Cbz-protected L-homoserine building block.
  • Supplied as a white to off-white solid for easy handling.
  • Melting point around 100-101 °C for characterization.
  • Available in small laboratory packaging sizes for research use.