Learn More
Medchemexpress LLC α-N-carbobenzoxy-L-lysine thiobenzyl ester monohydrochloride | 69861-89-8 | MFCD00058089 | 99.7% | C21H27ClN2O3S | 10MG
SDP
Catalog No. 5000260926
Encompass_Preferred
Z-LYS-SBZL monohydrochloride is the monohydrochloride salt of a carbobenzoxy-protected lysine thiobenzyl ester. Supplied as a solid research reagent, it is used in biochemical assays, protease and esterase activity studies, and peptide synthesis workflows where a protected lysine derivative is required.
  • High purity suitable for analytical and preparative work
  • Available in small-scale pack sizes for research use
  • Stable when stored sealed and away from moisture
  • Soluble in common organic solvents for reaction compatibility
  • Useful as a substrate or intermediate in protease and peptide studies
Catalog No. 50-002-60926 Supplier Medchemexpress LLC Supplier No. HY14144710MG
Z-LYS-SBZL monohydrochloride is the monohydrochloride salt of a carbobenzoxy-protected lysine thiobenzyl ester. Supplied as a solid research reagent, it is used in biochemical assays, protease and esterase activity studies, and peptide synthesis workflows where a protected lysine derivative is required.
  • High purity suitable for analytical and preparative work
  • Available in small-scale pack sizes for research use
  • Stable when stored sealed and away from moisture
  • Soluble in common organic solvents for reaction compatibility
  • Useful as a substrate or intermediate in protease and peptide studies