Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-phenylalanyl-L-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. C11701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C11701G Supplier TCI America Supplier No. C11701G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High purity biochemical reagent for research applications.

Chemical Identifiers

Molecular Formula C26H26N2O5
Molecular Weight (g/mol) 446.503
MDL Number MFCD00191139
InChI Key JNRHNGGTJOBXHL-GOTSBHOMSA-N
Synonym N-Cbz-L-phenylalanyl-L-phenylalanine, Z-Phe-Phe-OH
PubChem CID 114619
IUPAC Name (2S)-3-phenyl-2-[[(2S)-3-phenyl-2-(phenylmethoxycarbonylamino)propanoyl]amino]propanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)NC(CC2=CC=CC=C2)C(=O)O)NC(=O)OCC3=CC=CC=C3

Specifications

Melting Point 159°C
Color White
Quantity 1 g
Formula Weight 446.5
Percent Purity ≥98.0% (T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-phenylalanyl-L-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.