Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-proline 98.0+%, TCI America™
SDP

Catalog No. C071325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C071325G 25 g
C07135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C071325G Supplier TCI America Supplier No. C071325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1148-11-4
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD00003170
InChI Key JXGVXCZADZNAMJ-NSHDSACASA-N
Synonym z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline
PubChem CID 101987
IUPAC Name (2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 25 g
Formula Weight 249.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-proline
RTECSNumber UY0745000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.