Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-serine beta-Lactone 98.0+%, TCI America™
SDP

Catalog No. C15751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15751G Supplier TCI America Supplier No. C15751G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26054-60-4
Molecular Formula C11H11NO4
Molecular Weight (g/mol) 221.212
MDL Number MFCD01318309
InChI Key CWFZPRQDHIUBDO-VIFPVBQESA-N
Synonym (S)-alpha-Carbobenzoxyamino-beta-propiolactone, N-Cbz-L-serine beta-Lactone
PubChem CID 489182
IUPAC Name benzyl N-[(3S)-2-oxooxetan-3-yl]carbamate
SMILES C1C(C(=O)O1)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 1 g
Formula Weight 221.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-serine beta-Lactone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.