Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-S-phenyl-L-cysteine 98.0+%, TCI America™
SDP

Catalog No. C218025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C218025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C218025G Supplier TCI America Supplier No. C218025G

Chemical Identifiers

CAS 159453-24-4
Molecular Formula C17H17NO4S
Molecular Weight (g/mol) 331.386
MDL Number MFCD00671699
InChI Key ISBOGFMUFMJWEP-HNNXBMFYSA-N
Synonym cbz-s-phenyl-l-cysteine, z-cys ph-oh, n-z-s-phenyl-l-cysteine, n-cbz-s-phenyl-l-cysteine, n-carbobenzoxy-s-phenyl-l-cysteine, n-carbobenzyloxy-s-phenyl-l-cysteine, cbz-s-phenyl-l-cys-oh, r-2-benzyloxy carbonyl amino-3-phenylthio propanoic acid, n-benzyloxycarbonyl-s-phenyl-l-cysteine, n-carbobenzoxy-s-phenylcysteine
PubChem CID 2733947
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)-3-phenylsulfanylpropanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CSC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 25 g
Formula Weight 331.39
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-S-phenyl-L-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.