Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxyglycine 4-Nitrophenyl Ester 98.0+%, TCI America™
SDP

Catalog No. C03331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C03331G Supplier TCI America Supplier No. C03331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1738-86-9
Molecular Formula C16H14N2O6
Molecular Weight (g/mol) 330.296
MDL Number MFCD00024663
InChI Key LHFNPUGRSYOPLF-UHFFFAOYSA-N
Synonym N-Cbz-glycine 4-Nitrophenyl Ester, Z-Gly-ONp
PubChem CID 15616
IUPAC Name (4-nitrophenyl) 2-(phenylmethoxycarbonylamino)acetate
SMILES C1=CC=C(C=C1)COC(=O)NCC(=O)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 1 g
Formula Weight 330.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxyglycine 4-Nitrophenyl Ester
RTECSNumber MB9139000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.