Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzyloxy-beta-alanine, 98%

Catalog No. p-4228492
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434810050 5 g Glass Bottle
AC434810250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434810050 Supplier Thermo Scientific Chemicals Supplier No. 434810050
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2304-94-1 | C11H13NO4 | 223.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2304-94-1
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00037292
InChI Key GEVGRLPYQJTKKS-UHFFFAOYSA-N
Synonym z-beta-ala-oh, carbobenzyloxy-beta-alanine, n-cbz-beta-alanine, cbz-beta-alanine, n-benzyloxycarbonyl-beta-alanine, n-cbz-b-alanine, 3-benzyloxy carbonyl amino propanoic acid, cbz-beta-ala-oh, n-carbobenzoxy-beta-alanine
PubChem CID 75313
IUPAC Name 3-(phenylmethoxycarbonylamino)propanoic acid
SMILES OC(=O)CCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 105.3°C to 107.2°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 223.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Carbobenzyloxy-β-alanine
Show More Show Less
EINECSNumber 218-967-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.