Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzyloxy-L-isoleucine, 99+%, Thermo Scientific™

Catalog No. AC275980250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC275980250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC275980250 Supplier Thermo Scientific Chemicals Supplier No. 275980250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3160-59-6
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
InChI Key JSHXJPFZKBRLFU-JQWIXIFHSA-N
Synonym cbz-l-isoleucine, n-cbz-l-isoleucine, z-l-isoleucine, z-ile-oh, n-carbobenzyloxy-l-isoleucine, n-carbobenzoxy-l-isoleucine, cbz-l-ile-oh, n-benzyloxycarbonyl-l-isoleucine, l-isoleucine, n-phenylmethoxy carbonyl, 2s,3s-2-benzyloxy carbonyl amino-3-methylpentanoic acid
PubChem CID 2724772
IUPAC Name (2S,3S)-3-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CCC(C)C(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Linear Formula C6H5CH2OC(O)nHCH(COOH)CH(CH3)C2H5
Quantity 25 g
Specific Rotation Condition +7° (20°C c=6,ethanol)
Specific Rotation (20°C, 589nm) +6° to +8° (c=6, EtOH)
Formula Weight 265.3
Percent Purity ≥99.0%
Physical Form Weak Solid
Chemical Name or Material N-Carbobenzyloxy-L-isoleucine, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.