Learn More
Medchemexpress LLC Nα-(Carbobenzyloxy)-L-lysine | 2212-75-1 | MFCD00038204 | 98% | 280.32 | 25 G
SDP
Catalog No. 5000366433
Encompass_Preferred
Z-Lys-OH is also known as Nα-(Carbobenzyloxy)-L-lysine, Nα-Cbz-L-lysine, or Nα-Z-L-Lysine. It is an educt for the convenient preparation of L-α-aminoadipic acid and is used in solution phase peptide synthesis. It has an assay of 98% and an optical activity of [α]20/D -13±2°, c = 2 in 0.2 M HCl. The melting point is 226-231 °C (dec.).
  • Application: Peptide synthesis
  • Intermediate: Educt for the convenient preparation of L-α-aminoadipic acid
Catalog No. 50-003-66433 Supplier Medchemexpress LLC Supplier No. 25GHYW009337
Z-Lys-OH is also known as Nα-(Carbobenzyloxy)-L-lysine, Nα-Cbz-L-lysine, or Nα-Z-L-Lysine. It is an educt for the convenient preparation of L-α-aminoadipic acid and is used in solution phase peptide synthesis. It has an assay of 98% and an optical activity of [α]20/D -13±2°, c = 2 in 0.2 M HCl. The melting point is 226-231 °C (dec.).
  • Application: Peptide synthesis
  • Intermediate: Educt for the convenient preparation of L-α-aminoadipic acid