Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals N-Carbobenzyloxy-L-valine, 99+%

Catalog No. AC275940250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275940250 25 g Glass bottle
AC275940050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC275940250 Supplier Thermo Scientific Chemicals Supplier No. 275940250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1149-26-4 | C13H17NO4 | 251.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1149-26-4
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00008922
InChI Key CANZBRDGRHNSGZ-NSHDSACASA-N
Synonym n-carbobenzyloxy-l-valine, n-cbz-l-valine, z-val-oh, cbz-l-valine, n-benzyloxycarbonyl-l-valine, n-carbobenzoxy-l-valine, z-l-val-oh, carbobenzoxy-l-valine, z-valine, carbobenzyloxy-l-valine
PubChem CID 726987
SMILES CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 58°C to 60°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH(NHCO2CH2C6H5)CO2H
Quantity 25 g
Beilstein 06, 1502
Specific Rotation -4
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
Formula Weight 251.28
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzyloxy-L-valine, 99+%
Show More Show Less
EINECSNumber 214-562-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.