Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nε-(carboxyethyl)lysine-d4 | MFCD30364298 | 98.0% | 222.27 g/mol | C9H14D4N2O4 | 1 MG
SDP

Catalog No. 5000284559
Encompass_Preferred

Nε-(Carboxyethyl)lysine-d4 is a deuterium-labeled (d4) isotopic analogue of Nε-(carboxyethyl)lysine used as an internal standard for quantitative mass spectrometry and tracer studies. Supplied in small milligram quantities for analytical calibration, it supports accurate measurement of advanced glycation endproducts; consult the certificate of analysis for lot-specific purity, storage, and handling.

  • Deuterium-labeled (d4) isotopic internal standard for mass spectrometry.
  • Supplied in small, ready-to-use quantities suitable for trace analysis.
  • Facilitates accurate quantitation of advanced glycation endproducts (AGEs).
  • Compatible with common sample preparation and LC-MS workflows.
  • Chemical formula C9H14D4N2O4 and molecular weight ~222.27 g/mol.
  • Refer to the certificate of analysis for purity and storage details.

Catalog No. 50-002-84559 Supplier Medchemexpress LLC Supplier No. HYW585874S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nε-(Carboxyethyl)lysine-d4 is a deuterium-labeled (d4) isotopic analogue of Nε-(carboxyethyl)lysine used as an internal standard for quantitative mass spectrometry and tracer studies. Supplied in small milligram quantities for analytical calibration, it supports accurate measurement of advanced glycation endproducts; consult the certificate of analysis for lot-specific purity, storage, and handling.

  • Deuterium-labeled (d4) isotopic internal standard for mass spectrometry.
  • Supplied in small, ready-to-use quantities suitable for trace analysis.
  • Facilitates accurate quantitation of advanced glycation endproducts (AGEs).
  • Compatible with common sample preparation and LC-MS workflows.
  • Chemical formula C9H14D4N2O4 and molecular weight ~222.27 g/mol.
  • Refer to the certificate of analysis for purity and storage details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.