Learn More
Medchemexpress LLC Nε-(carboxymethyl)-L-lysine | 5746-04-3 | MFCD04114278 | 99.7% | 204.22 g·mol⁻1 | C8H16N2O4 | 10 MG
SDP
Catalog No. 5000282020
Encompass_Preferred
Nε-(Carboxymethyl)-L-lysine is a post-translationally formed advanced glycation end-product (AGE) commonly used as a biochemical reagent and reference standard in glycation research and biomarker assays. It is supplied as a white to off-white solid with molecular formula C8H16N2O4 and CAS 5746-04-3, and is used for assay development, calibration, and mechanistic studies of protein glycation.
  • Acts as a biomarker for advanced glycation end-products in biological samples.
  • Suitable as a reference standard for assay calibration and validation.
  • Available in small, solid quantities for analytical applications.
  • Soluble in water with sonication, facilitating in vitro preparations.
  • Stable when stored as a powder at -20 °C, enabling long-term storage.
  • Provided with supporting documentation such as COA and SDS.
Catalog No. 50-002-82020 Supplier Medchemexpress LLC Supplier No. HYW28674310MG
Nε-(Carboxymethyl)-L-lysine is a post-translationally formed advanced glycation end-product (AGE) commonly used as a biochemical reagent and reference standard in glycation research and biomarker assays. It is supplied as a white to off-white solid with molecular formula C8H16N2O4 and CAS 5746-04-3, and is used for assay development, calibration, and mechanistic studies of protein glycation.
  • Acts as a biomarker for advanced glycation end-products in biological samples.
  • Suitable as a reference standard for assay calibration and validation.
  • Available in small, solid quantities for analytical applications.
  • Soluble in water with sonication, facilitating in vitro preparations.
  • Stable when stored as a powder at -20 °C, enabling long-term storage.
  • Provided with supporting documentation such as COA and SDS.