Learn More
Medchemexpress LLC N-α-CBZ-L-arginine 7-amido-4-methylcoumarin hydrochloride | 70375-22-3 | MFCD00039019 | 99.2% | 501.96 g/mol | C24H28ClN5O5 | 5 MG
SDP
Catalog No. 5000223072
Encompass_Preferred
Z-Arg-AMC hydrochloride (CAS 70375-22-3) is a fluorogenic peptide substrate that releases 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage, enabling fluorescence-based assays to measure protease activity such as trypsin and papain. The compound is supplied as a white to off-white solid with high purity and is typically stored frozen to preserve stability.
  • Suitable for measuring trypsin and papain activity in fluorescence assays (Ex/Em 390/480 nm; alternative 355/460 nm).
  • High purity (99.2%) for reliable assay results.
  • Releases AMC fluorophore for sensitive detection of proteolytic activity.
  • White to off-white solid, convenient for weighing and formulation.
  • Stable under sealed storage at -20 °C; protect from moisture and light.
Catalog No. 50-002-23072 Supplier Medchemexpress LLC Supplier No. HY1344355MG
Z-Arg-AMC hydrochloride (CAS 70375-22-3) is a fluorogenic peptide substrate that releases 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage, enabling fluorescence-based assays to measure protease activity such as trypsin and papain. The compound is supplied as a white to off-white solid with high purity and is typically stored frozen to preserve stability.
  • Suitable for measuring trypsin and papain activity in fluorescence assays (Ex/Em 390/480 nm; alternative 355/460 nm).
  • High purity (99.2%) for reliable assay results.
  • Releases AMC fluorophore for sensitive detection of proteolytic activity.
  • White to off-white solid, convenient for weighing and formulation.
  • Stable under sealed storage at -20 °C; protect from moisture and light.