Learn More
Medchemexpress LLC N-CBZ-Phe-Arg-AMC TFA | 99.8% | 726.70 | 25 MG
SDP
Catalog No. 5000425482
Encompass_Preferred
N-CBZ-Phe-Arg-AMC (Z-FR-AMC) TFA is a cathepsin substrate used in the assessment of the activity of lysosomal cathepsin enzymes. It serves as a fluorescent substrate for trypsin, papain, and cathepsin B, allowing for the detection of apparent protease activity inhibition by myoglobin and hemin via a reduced fluorescence signal.
  • Cathepsin substrate
  • Used to assess the activity of lysosomal cathepsin enzymes
  • Fluorescent substrate for trypsin, papain, and cathepsin B
  • Detects apparent protease activity inhibition by myoglobin and hemin via a reduced fluorescence signal
Catalog No. 50-004-25482 Supplier Medchemexpress LLC Supplier No. 25MGHYP1759B
N-CBZ-Phe-Arg-AMC (Z-FR-AMC) TFA is a cathepsin substrate used in the assessment of the activity of lysosomal cathepsin enzymes. It serves as a fluorescent substrate for trypsin, papain, and cathepsin B, allowing for the detection of apparent protease activity inhibition by myoglobin and hemin via a reduced fluorescence signal.
  • Cathepsin substrate
  • Used to assess the activity of lysosomal cathepsin enzymes
  • Fluorescent substrate for trypsin, papain, and cathepsin B
  • Detects apparent protease activity inhibition by myoglobin and hemin via a reduced fluorescence signal