Learn More
Medchemexpress LLC N#CC1=C(N)SC2=C1[C@]3(CCC2)C4=C(CCC3)C(C5=NC(N6[C@H](CNCC6)C)=NC=C5)=NO4 | 2937344-16-4 | 99.8% | 461.58 g/mol | C24H27N7OS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15372310MG
Encompass_Preferred
BI-2493 is a small-molecule pan-KRAS inhibitor supplied for research use. It is reported to be highly selective and orally active, and has demonstrated concentration-dependent inhibition of KRAS-mutant cell proliferation in vitro and tumor-suppressive activity in multiple xenograft models in vivo.
  • Highly selective, orally active pan-KRAS inhibitor.
  • Inhibits proliferation of KRAS mutant cancer cell lines in vitro.
  • Exhibits tumor-suppressive activity in mouse xenograft models.
  • Reported purity 99.79% and molecular weight 461.58 g/mol.
  • Available in multiple formats and quantities, including 10 mg and 10 mM solutions in DMSO.
  • Supplied for research use only; not for human use.
Catalog No. 50-002-87698