Learn More
Medchemexpress LLC N-(chloroacetyl)-2-methoxybenzamide | 554423-67-5 | 98.0% | C10H10ClNO3 | 1 G
SDP
Catalog No. 5000434535
Encompass_Preferred
N-(Chloroacetyl)-2-methoxybenzamide is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.
  • Stable as powder for 3 years at -20°C
  • Stable as powder for 2 years at 4°C
  • Stable in solvent for 6 months at -80°C
  • Stable in solvent for 1 month at -20°C
Catalog No. 50-004-34535 Supplier Medchemexpress LLC Supplier No. HY773901G
N-(Chloroacetyl)-2-methoxybenzamide is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.
  • Stable as powder for 3 years at -20°C
  • Stable as powder for 2 years at 4°C
  • Stable in solvent for 6 months at -80°C
  • Stable in solvent for 1 month at -20°C