Learn More
Medchemexpress LLC N-(Chloroacetyl)-2-methoxybenzamide | 554423-67-5 | 98.0% | C10H10ClNO3 | 500 MG
SDP
Catalog No. 5000434606
Encompass_Preferred
N-(Chloroacetyl)-2-methoxybenzamide is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only and is not sold to patients.
  • Drug intermediate
  • Intended for research use only
  • Not sold to patients
  • Solid appearance
  • Shipped at room temperature in continental US (may vary elsewhere)
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-34606 Supplier Medchemexpress LLC Supplier No. HY77390500MG
N-(Chloroacetyl)-2-methoxybenzamide is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only and is not sold to patients.
  • Drug intermediate
  • Intended for research use only
  • Not sold to patients
  • Solid appearance
  • Shipped at room temperature in continental US (may vary elsewhere)
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month