Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Chloroacetyl-DL-2-aminobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. C06441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C06441G Supplier TCI America Supplier No. C06441G

Chemical Identifiers

CAS 101072-54-2
Molecular Formula C6H10ClNO3
Molecular Weight (g/mol) 179.6
MDL Number MFCD00055800
InChI Key CLYGVTIFEIIGIX-UHFFFAOYSA-N
PubChem CID 307936
IUPAC Name 2-[(2-chloroacetyl)amino]butanoic acid
SMILES CCC(C(=O)O)NC(=O)CCl

Specifications

Melting Point 130°C
Color White
Quantity 1 g
Formula Weight 179.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloroacetyl-DL-2-aminobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.